accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1002-16-0

IUPAC Name: pentyl nitrate
Synonyms: amyl nitrate
Molecular Weight (g/mol): 133.15
Molecular Formula: C5H11NO3
InChi Key: HSNWZBCBUUSSQD-UHFFFAOYSA-N
SMILES: CCCCCO[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results