accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1002-28-4

IUPAC Name: hex-3-yn-1-ol
Synonyms: 3-hexyn-1-ol
Molecular Weight (g/mol): 98.15
Molecular Formula: C6H10O
InChi Key: ODEHKVYXWLXRRR-UHFFFAOYSA-N
SMILES: CCC#CCCO
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)