accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1002-88-6

IUPAC Name: λ²-cobalt(2+) dioctadecanoate
Synonyms: λ²-cobalt(2+) bis(n-octadecanoate)
Molecular Weight (g/mol): 625.89
Molecular Formula: C36H70CoO4
InChi Key: AMFIJXSMYBKJQV-UHFFFAOYSA-L
SMILES: [Co++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results