accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 100202-39-9

IUPAC Name: methyl azetidine-3-carboxylate hydrochloride
Synonyms: methyl azetidine-3-carboxylate hydrochloride
Molecular Weight (g/mol): 151.59
Molecular Formula: C5H10ClNO2
InChi Key: UOCWTLBPYROHEF-UHFFFAOYSA-N
SMILES: Cl.COC(=O)C1CNC1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)