CAS RN 10025-65-7
Platinum(II) chloride, 99.9% (metals basis), Pt 73% min
CAS: 10025-65-7 Molecular Formula: Cl2Pt Molecular Weight (g/mol): 265.98 MDL Number: MFCD00011181 InChI Key: CLSUSRZJUQMOHH-UHFFFAOYSA-L IUPAC Name: platinum(2+) dichloride SMILES: [Cl-].[Cl-].[Pt++]
Platinum(II) chloride, 98%
CAS: 10025-65-7 Molecular Formula: Cl2Pt Molecular Weight (g/mol): 265.98 MDL Number: MFCD00011181 InChI Key: CLSUSRZJUQMOHH-UHFFFAOYSA-L IUPAC Name: platinum(2+) dichloride SMILES: [Cl-].[Cl-].[Pt++]
Platinum(II) chloride, 73% Pt
CAS: 10025-65-7 Molecular Formula: Cl2Pt Molecular Weight (g/mol): 265.98 MDL Number: MFCD00011181 InChI Key: CLSUSRZJUQMOHH-UHFFFAOYSA-L Synonym: Platinous chloride IUPAC Name: platinum(2+) dichloride SMILES: [Cl-].[Cl-].[Pt++]
Platinum(II) chloride, Premion™, 99.99+% (metals basis), Pt 73% min
CAS: 10025-65-7 Molecular Formula: Cl2Pt Molecular Weight (g/mol): 265.98 MDL Number: MFCD00011181 InChI Key: CLSUSRZJUQMOHH-UHFFFAOYSA-L IUPAC Name: platinum(2+) dichloride SMILES: [Cl-].[Cl-].[Pt++]
VeriSpec™ Platinum Standard for ICP/MS 10 ppm in 2% HCl, Ricca Chemical
CAS: 10025-65-7 Molecular Formula: Cl2Pt Molecular Weight (g/mol): 265.98 MDL Number: MFCD00011181 InChI Key: CLSUSRZJUQMOHH-UHFFFAOYSA-L IUPAC Name: platinum(2+) dichloride SMILES: [Cl-].[Cl-].[Pt++]
VeriSpec™ Platinum Standard for ICP 1000 ppm in 10% HCl, Ricca Chemical
CAS: 10025-65-7 Molecular Formula: Cl2Pt Molecular Weight (g/mol): 265.98 MDL Number: MFCD00011181 InChI Key: CLSUSRZJUQMOHH-UHFFFAOYSA-L IUPAC Name: platinum(2+) dichloride SMILES: [Cl-].[Cl-].[Pt++]
VeriSpec™ Platinum Standard for ICP/MS 100 ppm in 2% HCl, Ricca Chemical
CAS: 10025-65-7 Molecular Formula: Cl2Pt Molecular Weight (g/mol): 265.98 MDL Number: MFCD00011181 InChI Key: CLSUSRZJUQMOHH-UHFFFAOYSA-L IUPAC Name: platinum(2+) dichloride SMILES: [Cl-].[Cl-].[Pt++]
VeriSpec™ Platinum (Pt) Standard for AAS 1000 ppm in 10% HCl, Ricca Chemical
CAS: 10025-65-7 Molecular Formula: Cl2Pt Molecular Weight (g/mol): 265.98 MDL Number: MFCD00011181 InChI Key: CLSUSRZJUQMOHH-UHFFFAOYSA-L IUPAC Name: platinum(2+) dichloride SMILES: [Cl-].[Cl-].[Pt++]
Platinum(II) Chloride 95.0+%, TCI America™
CAS: 10025-65-7 Molecular Formula: Cl2Pt Molecular Weight (g/mol): 265.98 MDL Number: MFCD00011181 InChI Key: CLSUSRZJUQMOHH-UHFFFAOYSA-L IUPAC Name: platinum(2+) dichloride SMILES: [Cl-].[Cl-].[Pt++]
Sigma Aldrich Platinum(II) chloride
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Platinum(II) chloride
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Platinum(II) Chloride
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.