accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClSeSeCl

CAS RN 10025-68-0

IUPAC Name: dichlorodiselenane
Synonyms: selenium monochloride
Molecular Weight (g/mol): 228.84
Molecular Formula: Cl2Se2
InChi Key: VIEXQFHKRAHTQS-UHFFFAOYSA-N
SMILES: Cl[Se][Se]Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results