accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10025-74-8

ClClClDy

CAS RN 10025-74-8

IUPAC Name: dysprosium(3+) trichloride
Synonyms: dysprosium(III) chloride
Molecular Weight (g/mol): 268.85
Molecular Formula: Cl3Dy
InChi Key: BOXVSFHSLKQLNZ-UHFFFAOYSA-K
SMILES: [Cl-].[Cl-].[Cl-].[Dy+3]
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)