accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10025-76-0

IUPAC Name: europium(3+) trichloride
Synonyms: europium(III) chloride
Molecular Weight (g/mol): 258.31
Molecular Formula: Cl3Eu
InChi Key: NNMXSTWQJRPBJZ-UHFFFAOYSA-K
SMILES: [Cl-].[Cl-].[Cl-].[Eu+3]
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (3)
  • (2)
Quantity 
  • (3)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)