CAS RN 10025-91-9
Antimony trichloride, ACS reagent
CAS: 10025-91-9 Molecular Formula: Cl3Sb Molecular Weight (g/mol): 228.11 MDL Number: MFCD00011212 InChI Key: FAPDDOBMIUGHIN-UHFFFAOYSA-K Synonym: antimony trichloride,trichlorostibine,antimony chloride,antimony iii chloride,stibine, trichloro,antimontrichlorid,antimonous chloride,butter of antimony,antimony butter,caustic antimony PubChem CID: 24814 ChEBI: CHEBI:74856 SMILES: [Cl-].[Cl-].[Cl-].[Sb+3]
Antimony(III) chloride, 99+%
CAS: 10025-91-9 Molecular Formula: Cl3Sb Molecular Weight (g/mol): 228.11 MDL Number: MFCD00011212 InChI Key: FAPDDOBMIUGHIN-UHFFFAOYSA-K Synonym: antimony trichloride,trichlorostibine,antimony chloride,antimony iii chloride,stibine, trichloro,antimontrichlorid,antimonous chloride,butter of antimony,antimony butter,caustic antimony PubChem CID: 24814 ChEBI: CHEBI:74856 SMILES: [Cl-].[Cl-].[Cl-].[Sb+3]
Antimony(III) chloride, ACS, 99.0% min
CAS: 10025-91-9 Molecular Formula: Cl3Sb Molecular Weight (g/mol): 228.11 MDL Number: MFCD00011212 InChI Key: FAPDDOBMIUGHIN-UHFFFAOYSA-K Synonym: antimony trichloride,trichlorostibine,antimony chloride,antimony iii chloride,stibine, trichloro,antimontrichlorid,antimonous chloride,butter of antimony,antimony butter,caustic antimony PubChem CID: 24814 ChEBI: CHEBI:74856 IUPAC Name: trichlorostibane SMILES: [Cl-].[Cl-].[Cl-].[Sb+3]
Antimony(III) chloride, 99.9% (metals basis)
CAS: 10025-91-9 Molecular Formula: Cl3Sb Molecular Weight (g/mol): 228.11 MDL Number: MFCD00011212 InChI Key: FAPDDOBMIUGHIN-UHFFFAOYSA-K Synonym: antimony trichloride,trichlorostibine,antimony chloride,antimony iii chloride,stibine, trichloro,antimontrichlorid,antimonous chloride,butter of antimony,antimony butter,caustic antimony PubChem CID: 24814 ChEBI: CHEBI:74856 IUPAC Name: trichlorostibane SMILES: [Cl-].[Cl-].[Cl-].[Sb+3]
Antimony Trichloride TS, (U.S.P. Test Solution), Spectrum™ Chemical
CAS: 10025-91-9 Molecular Formula: Cl3Sb Molecular Weight (g/mol): 228.11 MDL Number: MFCD00011212 InChI Key: FAPDDOBMIUGHIN-UHFFFAOYSA-K IUPAC Name: antimony(3+) trichloride SMILES: [Cl-].[Cl-].[Cl-].[Sb+3]
Antimony trichloride, 99.5%
CAS: 10025-91-9 Molecular Formula: Cl3Sb Molecular Weight (g/mol): 228.11 MDL Number: MFCD00011212 InChI Key: FAPDDOBMIUGHIN-UHFFFAOYSA-K Synonym: antimony trichloride,trichlorostibine,antimony chloride,antimony iii chloride,stibine, trichloro,antimontrichlorid,antimonous chloride,butter of antimony,antimony butter,caustic antimony PubChem CID: 24814 ChEBI: CHEBI:74856 SMILES: [Cl-].[Cl-].[Cl-].[Sb+3]
Antimony(III) chloride, ultra dry, 99.999% (metals basis)
CAS: 10025-91-9 Molecular Formula: Cl3Sb Molecular Weight (g/mol): 228.11 MDL Number: MFCD00011212 InChI Key: FAPDDOBMIUGHIN-UHFFFAOYSA-K Synonym: antimony trichloride,trichlorostibine,antimony chloride,antimony iii chloride,stibine, trichloro,antimontrichlorid,antimonous chloride,butter of antimony,antimony butter,caustic antimony PubChem CID: 24814 ChEBI: CHEBI:74856 SMILES: [Cl-].[Cl-].[Cl-].[Sb+3]
Antimony(III) Chloride, ACS, For Analysis, EMSURE™, MilliporeSigma™
CAS: 10025-91-9 Molecular Formula: Cl3Sb Molecular Weight (g/mol): 228.11 MDL Number: MFCD00011212 InChI Key: FAPDDOBMIUGHIN-UHFFFAOYSA-K Synonym: antimony trichloride,trichlorostibine,antimony chloride,antimony iii chloride,stibine, trichloro,antimontrichlorid,antimonous chloride,butter of antimony,antimony butter,caustic antimony PubChem CID: 24814 ChEBI: CHEBI:74856 IUPAC Name: antimony(3+) trichloride SMILES: [Cl-].[Cl-].[Cl-].[Sb+3]
Antimony(III) chloride, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
CAS: 10025-91-9 Molecular Formula: Cl3Sb Molecular Weight (g/mol): 228.11 MDL Number: MFCD00011212 InChI Key: FAPDDOBMIUGHIN-UHFFFAOYSA-K Synonym: antimony trichloride,trichlorostibine,antimony chloride,antimony iii chloride,stibine, trichloro,antimontrichlorid,antimonous chloride,butter of antimony,antimony butter,caustic antimony PubChem CID: 24814 ChEBI: CHEBI:74856 SMILES: [Cl-].[Cl-].[Cl-].[Sb+3]
VeriSpec™ Antimony (Sb) Standard for AAS 1000 ppm in 20% HCl, Ricca Chemical
CAS: 10025-91-9 Molecular Formula: Cl3Sb Molecular Weight (g/mol): 228.11 MDL Number: MFCD00011212 InChI Key: FAPDDOBMIUGHIN-UHFFFAOYSA-K PubChem CID: 24814 ChEBI: CHEBI:74856 IUPAC Name: antimony(3+) trichloride SMILES: [Cl-].[Cl-].[Cl-].[Sb+3]
Antimony(Iii) Chloride, puriss. p.a.ACS Reagent, ≥99.0% (RT), Solstice
CAS: 10025-91-9 Molecular Formula: Cl3Sb Molecular Weight (g/mol): 228.11 MDL Number: MFCD00011212 InChI Key: FAPDDOBMIUGHIN-UHFFFAOYSA-K Synonym: antimony trichloride,trichlorostibine,antimony chloride,antimony iii chloride,stibine, trichloro,antimontrichlorid,antimonous chloride,butter of antimony,antimony butter,caustic antimony PubChem CID: 24814 ChEBI: CHEBI:74856 IUPAC Name: trichlorostibane SMILES: [Cl-].[Cl-].[Cl-].[Sb+3]
Sigma Aldrich Antimony(III) chloride
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Antimony, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical™
CAS: 10025-91-9 Molecular Formula: Cl3Sb Molecular Weight (g/mol): 228.11 MDL Number: MFCD00011212 InChI Key: FAPDDOBMIUGHIN-UHFFFAOYSA-K PubChem CID: 24814 ChEBI: CHEBI:74856 IUPAC Name: antimony(3+) trichloride SMILES: [Cl-].[Cl-].[Cl-].[Sb+3]
- 1
- 2