accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10025-98-6

IUPAC Name: dipotassium tetrachloropalladiumdiuide
Synonyms: dipotassium tetrachloropalladate
Molecular Weight (g/mol): 330.45
Molecular Formula: Cl4H4K2Pd
InChi Key: OANHYYMESYMMDY-UHFFFAOYSA-N
SMILES: Cl.Cl.Cl.Cl.[K+].[K+].[Pd--]
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)