accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10026-03-6

IUPAC Name: tetrachloro-λ⁴-selane
Synonyms: selenium tetrachloride
Molecular Weight (g/mol): 220.77
Molecular Formula: Cl4Se
InChi Key: LNBXMNQCXXEHFT-UHFFFAOYSA-N
SMILES: Cl[Se](Cl)(Cl)Cl
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)