accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10026-07-0

IUPAC Name: tetrachloro-λ⁴-tellane
Synonyms: tellurium tetrachloride
Molecular Weight (g/mol): 269.4
Molecular Formula: Cl4Te
InChi Key: SWLJJEFSPJCUBD-UHFFFAOYSA-N
SMILES: Cl[Te](Cl)(Cl)Cl
Molecular Weight (g/mol) 
  • (9)
  • (1)
Grade 
  • (5)
Quantity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results