accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10026-18-3

IUPAC Name: cobalt(3+) trifluoride
Synonyms: cobalt(III) fluoride
Molecular Weight (g/mol): 115.93
Molecular Formula: CoF3
InChi Key: WZJQNLGQTOCWDS-UHFFFAOYSA-K
SMILES: [F-].[F-].[F-].[Co+3]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results