accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10027-07-3

IUPAC Name: octanedioyl dichloride
Synonyms: octanedioyl dichloride
Molecular Weight (g/mol): 211.08
Molecular Formula: C8H12Cl2O2
InChi Key: PUIBKAHUQOOLSW-UHFFFAOYSA-N
SMILES: ClC(=O)CCCCCCC(Cl)=O
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)