accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CH3CCH3H3CO

CAS RN 1002727-88-9

IUPAC Name: 2-(3,4-dihydro-2H-1-benzopyran-6-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(3,4-dihydro-2H-1-benzopyran-6-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 260.14
Molecular Formula: C15H21BO3
InChi Key: NDSHAELUPJMEBM-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC2=C(OCCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results