accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1002727-89-0

IUPAC Name: 4-(phenoxymethyl)benzene-1-sulfonyl chloride
Synonyms: 4-(phenoxymethyl)benzenesulfonyl chloride
Molecular Weight (g/mol): 282.74
Molecular Formula: C13H11ClO3S
InChi Key: ITLWVYCIZYWFNW-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=C(COC2=CC=CC=C2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results