accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1002727-90-3

IUPAC Name: 4-[(thiophen-2-yl)methyl]benzoic acid
Synonyms: 4-(thiophen-2-ylmethyl)benzoic acid
Molecular Weight (g/mol): 218.27
Molecular Formula: C12H10O2S
InChi Key: OAIBKSRZDOMJKZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(CC2=CC=CS2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results