accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 100277-27-8

IUPAC Name: (thian-4-yl)methanol
Synonyms: thian-4-ylmethanol
Molecular Weight (g/mol): 132.22
Molecular Formula: C6H12OS
InChi Key: LMVCLDAMMNHILO-UHFFFAOYSA-N
SMILES: OCC1CCSCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results