accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFNi

CAS RN 10028-18-9

IUPAC Name: nickel(2+) difluoride
Synonyms: nickel(II) fluoride
Molecular Weight (g/mol): 96.69
Molecular Formula: F2Ni
InChi Key: DBJLJFTWODWSOF-UHFFFAOYSA-L
SMILES: [F-].[F-].[Ni++]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results