accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH2OOSOOOFe

CAS RN 10028-21-4

IUPAC Name: λ²-iron(2+) dihydrate sulfate
Synonyms: λ²-iron(2+) dihydrate sulfate
Molecular Weight (g/mol): 187.93
Molecular Formula: FeH4O6S
InChi Key: KFJBRJOPXNCARV-UHFFFAOYSA-L
SMILES: O.O.[Fe++].[O-]S([O-])(=O)=O
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results