accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 100299-08-9

IUPAC Name: potassium 5-{9-methyl-4-oxo-4H-pyrido[1,2-a]pyrimidin-3-yl}-1H-1,2,3,4-tetrazol-1-ide
Synonyms: pemirolast
Molecular Weight (g/mol): 266.31
Molecular Formula: C10H7KN6O
InChi Key: NMMVKSMGBDRONO-UHFFFAOYSA-N
SMILES: [K+].CC1=CC=CN2C(=O)C(=CN=C12)C1=NN=N[N-]1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results