accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1003-03-8

IUPAC Name: cyclopentanamine
Synonyms: cyclopentanamine
Molecular Weight (g/mol): 85.15
Molecular Formula: C5H11N
InChi Key: NISGSNTVMOOSJQ-UHFFFAOYSA-N
SMILES: NC1CCCC1
Boiling Point 
  • (6)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (4)
  • (4)
Quantity 
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (6)
  • (2)
  • (2)
  • (2)
  • (4)
  • (4)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)