accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1003-21-0

IUPAC Name: 5-bromo-1-methyl-1H-imidazole
Synonyms: 5-bromo-1-methylimidazole
Molecular Weight (g/mol): 161
Molecular Formula: C4H5BrN2
InChi Key: HATLLUIOEIXWGD-UHFFFAOYSA-N
SMILES: CN1C=NC=C1Br
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)