accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10030-67-8

IUPAC Name: 2-{[4-hydroxy-2,5-bis(hydroxymethyl)-2-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxolan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol dihydrate
Synonyms: 2-{[4-hydroxy-2,5-bis(hydroxymethyl)-2-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxolan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol dihydrate
Molecular Weight (g/mol): 540.47
Molecular Formula: C18H36O18
InChi Key: LNVIPYYEBMNJIL-UHFFFAOYNA-N
SMILES: O.O.OCC1OC(CO)(OC2OC(CO)C(O)C(O)C2O)C(OC2OC(CO)C(O)C(O)C2O)C1O
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results