accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10031-20-6

IUPAC Name: manganese(2+) tetrahydrate dibromide
Synonyms: manganese(2+) tetrahydrate dibromide
Molecular Weight (g/mol): 286.81
Molecular Formula: Br2H8MnO4
InChi Key: HHDPJRSXPOGIOP-UHFFFAOYSA-L
SMILES: O.O.O.O.[Mn++].[Br-].[Br-]
Percent Purity 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (6)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)