accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10031-26-2

IUPAC Name: iron(3+) tribromide
Synonyms: iron(III) bromide
Molecular Weight (g/mol): 295.56
Molecular Formula: Br3Fe
InChi Key: FEONEKOZSGPOFN-UHFFFAOYSA-K
SMILES: [Fe+3].[Br-].[Br-].[Br-]
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)