CAS RN 10031-30-8
IUPAC Name:
calcium hydrate diphosphate
Synonyms:
calcium hydrate diphosphate
Molecular Weight (g/mol):
248.04
Molecular Formula:
CaH2O9P2
InChi Key:
ZBZJARSYCHAEND-UHFFFAOYSA-H
SMILES:
O.[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
1
–
2
of
2
results
1
Promotions Available
Calcium Phosphate, Monobasic, Monohydrate, Crystal, 15.9-17.7%, Spectrum™ Chemical
Calcium Phosphate, Monobasic, Monohydrate, Crystal, 15.9-17.7%, Spectrum™ Chemical
CAS: 10031-30-8
2
Promotions Available
Calcium Phosphate, Monobasic, Monohydrate, Powder, FCC, 15.9-17.7%, Spectrum™ Chemical
Calcium Phosphate, Monobasic, Monohydrate, Powder, FCC, 15.9-17.7%, Spectrum™ Chemical
CAS: 10031-30-8 Molecular Formula: CaH2O9P2 Molecular Weight (g/mol): 248.04 InChI Key: ZBZJARSYCHAEND-UHFFFAOYSA-H IUPAC Name: calcium hydrate diphosphate SMILES: O.[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O