accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10031-51-3

IUPAC Name: erbium(3+) pentahydrate trinitrate
Synonyms: erbium(3+) pentahydrate trinitrate
Molecular Weight (g/mol): 443.35
Molecular Formula: ErH10N3O14
InChi Key: LWHHUEHWVBVASY-UHFFFAOYSA-N
SMILES: O.O.O.O.O.[Er+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Grade 
  • (8)
  • (2)
  • (2)
Percent Purity 
  • (4)
  • (2)
  • (2)
Quantity 
  • (4)
  • (4)
Physical Form 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (2)
  • (6)
  • (8)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (2)