accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10031-90-0

IUPAC Name: ethyl 3-(furan-2-yl)propanoate
Synonyms: ethyl 3-(furan-2-yl)propanoate
Molecular Weight (g/mol): 168.19
Molecular Formula: C9H12O3
InChi Key: OWIWZQQFSTZZIG-UHFFFAOYSA-N
SMILES: CCOC(=O)CCC1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results