accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO

CAS RN 10031-93-3

IUPAC Name: ethyl 4-phenylbutanoate
Synonyms: ethyl 4-phenylbutanoate
Molecular Weight (g/mol): 192.26
Molecular Formula: C12H16O2
InChi Key: GGFNXKFGVQQNRV-UHFFFAOYSA-N
SMILES: CCOC(=O)CCCC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results