accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CFNF

CAS RN 1003298-73-4

IUPAC Name: 2,6-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Synonyms: 2,6-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Molecular Weight (g/mol): 265.07
Molecular Formula: C13H14BF2NO2
InChi Key: XYMVJTWEERDWFY-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(F)=C(C#N)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results