accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CClOHOCH3

CAS RN 1003298-84-7

IUPAC Name: 2-chloro-6-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Synonyms: 2-chloro-6-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Molecular Weight (g/mol): 284.54
Molecular Formula: C13H18BClO4
InChi Key: XBEJPOZGAPQFPL-UHFFFAOYSA-N
SMILES: COC1=C(O)C(Cl)=CC(=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results