accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3COOOOCH3ONH2OCH3OOCH3O

CAS RN 10034-20-5

IUPAC Name: [3,4,6-tris(acetyloxy)-5-aminooxan-2-yl]methyl acetate hydrochloridyl
Synonyms: [3,4,6-tris(acetyloxy)-5-aminooxan-2-yl]methyl acetate hydrochloridyl
Molecular Weight (g/mol): 382.77
Molecular Formula: C14H21ClNO9
InChi Key: NYYNGCPGIUHVAL-UHFFFAOYNA-N
SMILES: [Cl].CC(=O)OCC1OC(OC(C)=O)C(N)C(OC(C)=O)C1OC(C)=O
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)