accessibility menu, dialog, popup

UserName

Viewing profile as user:
MgOClOOOOClOOO

CAS RN 10034-81-8

IUPAC Name: magnesium(2+) diperchlorate
Synonyms: magnesium perchlorate
Molecular Weight (g/mol): 223.2
Molecular Formula: Cl2MgO8
InChi Key: MPCRDALPQLDDFX-UHFFFAOYSA-L
SMILES: [Mg++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Grade 
  • (1)
  • (4)
  • (4)
Molecular Weight (g/mol) 
  • (3)
  • (4)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
Physical Form 
  • (2)
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (4)
  • (4)
  • (1)
  • (4)
  • (4)
  • (3)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (4)