accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10034-85-2

IUPAC Name: hydrogen iodide
Synonyms: hydrogen iodide|H-I|hydroiodic acid|hydrogen iodide (HI)|iodinane|hydriodic acid, solution|hydrogen iodide, anhydrous|anhydrous hydrogen iodide|jodwasserstoff|hydrogen iodide solution
Molecular Weight (g/mol): 127.91
Molecular Formula: HI
InChi Key: XMBWDFGMSWQBCA-UHFFFAOYSA-N
SMILES: I
Quantity 
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (17)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)