CAS RN 10035-10-6
Hydrobromic acid, ACS, 47.0-49.0%
CAS: 10035-10-6 Molecular Formula: BrH Molecular Weight (g/mol): 80.91 MDL Number: MFCD00011323 InChI Key: CPELXLSAUQHCOX-UHFFFAOYSA-N Synonym: hydrogen bromide,hydrobromic acid,bromwasserstoff,broomwaterstof,bromowodor,acido bromidrico,acide bromhydrique,anhydrous hydrobromic acid,bromowodor polish,hydrobromide PubChem CID: 260 ChEBI: CHEBI:47266 SMILES: Br
Hydrobromic Acid (OPTIMA™), Fisher Chemical™
CAS: 10035-10-6 Molecular Formula: BrH Molecular Weight (g/mol): 80.91 MDL Number: MFCD00011323 InChI Key: CPELXLSAUQHCOX-UHFFFAOYSA-N Synonym: hydrogen bromide,hydrobromic acid,bromwasserstoff,broomwaterstof,bromowodor,acido bromidrico,acide bromhydrique,anhydrous hydrobromic acid,bromowodor polish,hydrobromide PubChem CID: 260 ChEBI: CHEBI:47266 IUPAC Name: hydrogen bromide SMILES: Br
Hydrobromic acid, 99.9999% (metals basis), 48% w/w aq. soln.
CAS: 10035-10-6 Molecular Formula: BrH Molecular Weight (g/mol): 80.91 MDL Number: MFCD00011323 InChI Key: CPELXLSAUQHCOX-UHFFFAOYSA-N Synonym: hydrogen bromide,hydrobromic acid,bromwasserstoff,broomwaterstof,bromowodor,acido bromidrico,acide bromhydrique,anhydrous hydrobromic acid,bromowodor polish,hydrobromide PubChem CID: 260 ChEBI: CHEBI:47266 SMILES: Br
Hydrogen bromide, pure, 33 wt% solution in glacial acetic acid
CAS: 10035-10-6 Molecular Formula: BrH Molecular Weight (g/mol): 80.91 MDL Number: MFCD00011323 InChI Key: CPELXLSAUQHCOX-UHFFFAOYSA-N Synonym: hydrogen bromide,hydrobromic acid,bromwasserstoff,broomwaterstof,bromowodor,acido bromidrico,acide bromhydrique,anhydrous hydrobromic acid,bromowodor polish,hydrobromide PubChem CID: 260 ChEBI: CHEBI:47266 SMILES: Br
Hydrobromic acid, pure, ca. 48 wt% solution in water
CAS: 10035-10-6 Molecular Formula: BrH Molecular Weight (g/mol): 80.91 MDL Number: MFCD00011323 InChI Key: CPELXLSAUQHCOX-UHFFFAOYSA-N Synonym: hydrogen bromide,hydrobromic acid,bromwasserstoff,broomwaterstof,bromowodor,acido bromidrico,acide bromhydrique,anhydrous hydrobromic acid,bromowodor polish,hydrobromide PubChem CID: 260 ChEBI: CHEBI:47266 IUPAC Name: bromane SMILES: Br
Hydrogen bromide, pure, 33 wt.% sol. in glacial acetic acid, AcroSeal™
CAS: 10035-10-6 Molecular Formula: BrH Molecular Weight (g/mol): 80.91 MDL Number: MFCD00011323 InChI Key: CPELXLSAUQHCOX-UHFFFAOYSA-N Synonym: hydrogen bromide,hydrobromic acid,bromwasserstoff,broomwaterstof,bromowodor,acido bromidrico,acide bromhydrique,anhydrous hydrobromic acid,bromowodor polish,hydrobromide PubChem CID: 260 ChEBI: CHEBI:47266 SMILES: Br
Hydrobromic acid, ACS reagent, ca. 48% solution in water
CAS: 10035-10-6 Molecular Formula: BrH Molecular Weight (g/mol): 80.91 MDL Number: MFCD00011323 InChI Key: CPELXLSAUQHCOX-UHFFFAOYSA-N Synonym: hydrogen bromide,hydrobromic acid,bromwasserstoff,broomwaterstof,bromowodor,acido bromidrico,acide bromhydrique,anhydrous hydrobromic acid,bromowodor polish,hydrobromide PubChem CID: 260 ChEBI: CHEBI:47266 IUPAC Name: hydrogen bromide SMILES: Br
Hydrobromic acid, 48% w/w aq. soln.
CAS: 10035-10-6 Molecular Formula: BrH Molecular Weight (g/mol): 80.91 MDL Number: MFCD00011323 InChI Key: CPELXLSAUQHCOX-UHFFFAOYSA-N Synonym: hydrogen bromide,hydrobromic acid,bromwasserstoff,broomwaterstof,bromowodor,acido bromidrico,acide bromhydrique,anhydrous hydrobromic acid,bromowodor polish,hydrobromide PubChem CID: 260 ChEBI: CHEBI:47266 IUPAC Name: hydrogen bromide SMILES: Br
Hydrobromic Acid, purum p.a., Solstice
CAS: 10035-10-6 Molecular Formula: BrH Molecular Weight (g/mol): 80.91 MDL Number: MFCD00011323 InChI Key: CPELXLSAUQHCOX-UHFFFAOYSA-N Synonym: hydrogen bromide,hydrobromic acid,bromwasserstoff,broomwaterstof,bromowodor,acido bromidrico,acide bromhydrique,anhydrous hydrobromic acid,bromowodor polish,hydrobromide PubChem CID: 260 ChEBI: CHEBI:47266 IUPAC Name: bromane SMILES: Br
Hydrobromic acid, for analysis, ca. 48% solution in water
CAS: 10035-10-6 Molecular Formula: BrH Molecular Weight (g/mol): 80.91 MDL Number: MFCD00011323 InChI Key: CPELXLSAUQHCOX-UHFFFAOYSA-N Synonym: hydrogen bromide,hydrobromic acid,bromwasserstoff,broomwaterstof,bromowodor,acido bromidrico,acide bromhydrique,anhydrous hydrobromic acid,bromowodor polish,hydrobromide PubChem CID: 260 ChEBI: CHEBI:47266 IUPAC Name: hydrogen bromide SMILES: Br
Hydrobromic Acid, 47.0%, Suprapur™, MilliporeSigma™
CAS: 10035-10-6 Molecular Formula: BrH Molecular Weight (g/mol): 80.91 MDL Number: MFCD00011323 InChI Key: CPELXLSAUQHCOX-UHFFFAOYSA-N Synonym: Hydrogen bromide solution IUPAC Name: hydrogen bromide SMILES: Br
Hydrobromic Acid, 47% to 49%, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
CAS: 10035-10-6 Molecular Formula: BrH Molecular Weight (g/mol): 80.91 MDL Number: MFCD00011323 InChI Key: CPELXLSAUQHCOX-UHFFFAOYSA-N Synonym: hydrogen bromide,hydrobromic acid,bromwasserstoff,broomwaterstof,bromowodor,acido bromidrico,acide bromhydrique,anhydrous hydrobromic acid,bromowodor polish,hydrobromide PubChem CID: 260 ChEBI: CHEBI:47266 SMILES: Br
- 1
- 2