accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1003711-43-0

IUPAC Name: 6-bromo-5-methylpyridin-3-ol
Synonyms: 6-bromo-5-methylpyridin-3-ol
Molecular Weight (g/mol): 188.02
Molecular Formula: C6H6BrNO
InChi Key: KCCYUVIIKCPVDH-UHFFFAOYSA-N
SMILES: CC1=CC(O)=CN=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results