accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CBHOOH

CAS RN 100379-00-8

IUPAC Name: (2,6-dimethylphenyl)boronic acid
Synonyms: 2,6-dimethylphenylboronic acid
Molecular Weight (g/mol): 149.98
Molecular Formula: C8H11BO2
InChi Key: ZXDTWWZIHJEZOG-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=C1B(O)O
Molecular Weight (g/mol) 
  • (8)
  • (2)
Quantity 
  • (3)
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (8)
  • (2)
  • (3)
  • (3)
  • (4)
  • (2)
  • (3)
  • (3)
  • (2)