accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1003845-08-6

IUPAC Name: 2-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine
Synonyms: 2-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine
Molecular Weight (g/mol): 240.49
Molecular Formula: C10H14BClN2O2
InChi Key: VLAPDEKXZLRRKV-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CN=C(Cl)N=C1
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)