accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1004-00-8

IUPAC Name: 2-amino-4-chlorobenzene-1-thiol
Synonyms: 2-amino-4-chlorobenzenethiol
Molecular Weight (g/mol): 159.63
Molecular Formula: C6H6ClNS
InChi Key: NGIRMPARLVGMPX-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=CC=C1S
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)