accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10042-59-8

IUPAC Name: 2-propylheptan-1-ol
Synonyms: 1-heptanol, 2-propyl- propylheptyl alcohol
Molecular Weight (g/mol): 158.29
Molecular Formula: C10H22O
InChi Key: YLQLIQIAXYRMDL-UHFFFAOYNA-N
SMILES: CCCCCC(CO)CCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results