accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10043-27-3

IUPAC Name: terbium(3+) trinitrate
Synonyms: terbium(III) nitrate
Molecular Weight (g/mol): 344.94
Molecular Formula: N3O9Tb
InChi Key: YJVUGDIORBKPLC-UHFFFAOYSA-N
SMILES: [Tb+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Grade 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)