CAS RN 10043-35-3
Boric Acid (Powder/Certified ACS), Fisher Chemical™
CAS: 10043-35-3 Molecular Formula: BH3O3 Molecular Weight (g/mol): 61.83 MDL Number: MFCD00011337 InChI Key: KGBXLFKZBHKPEV-UHFFFAOYSA-N Synonym: orthoboric acid,boracic acid,borofax,boron hydroxide,boron trihydroxide,three elephant,basilit b,trihydroxyborone,h3bo3,flea prufe PubChem CID: 7628 ChEBI: CHEBI:33118 IUPAC Name: boric acid SMILES: OB(O)O
Boric Acid (Crystalline/Certified ACS), Fisher Chemical™
CAS: 10043-35-3 Molecular Formula: BH3O3 Molecular Weight (g/mol): 61.83 MDL Number: MFCD00011337 InChI Key: KGBXLFKZBHKPEV-UHFFFAOYSA-N Synonym: orthoboric acid,boracic acid,borofax,boron hydroxide,boron trihydroxide,three elephant,basilit b,trihydroxyborone,h3bo3,flea prufe PubChem CID: 7628 ChEBI: CHEBI:33118 IUPAC Name: boric acid SMILES: OB(O)O
Corning™ Molecular Biology Reagents
Used in molecular biology, nucleic acid purification applications
TBE Buffer, Tris-Borate-EDTA, 10X Solution, Electrophoresis, Fisher BioReagents™
CAS: 10043-35-3 Synonym: TBE
Boric acid, 99.5%, for molecular biology, DNAse, RNAse and Protease free
CAS: 10043-35-3 Molecular Formula: BH3O3 Molecular Weight (g/mol): 61.83 MDL Number: MFCD00011337 InChI Key: KGBXLFKZBHKPEV-UHFFFAOYSA-N Synonym: orthoboric acid,boracic acid,borofax,boron hydroxide,boron trihydroxide,three elephant,basilit b,trihydroxyborone,h3bo3,flea prufe PubChem CID: 7628 ChEBI: CHEBI:33118 IUPAC Name: boric acid SMILES: OB(O)O
Boric acid, 99.6%, ACS reagent
CAS: 10043-35-3 Molecular Formula: BH3O3 Molecular Weight (g/mol): 61.83 MDL Number: MFCD00011337 InChI Key: KGBXLFKZBHKPEV-UHFFFAOYSA-N Synonym: orthoboric acid,boracic acid,borofax,boron hydroxide,boron trihydroxide,three elephant,basilit b,trihydroxyborone,h3bo3,flea prufe PubChem CID: 7628 ChEBI: CHEBI:33118 IUPAC Name: boric acid SMILES: OB(O)O
Boric acid, 99+%
CAS: 10043-35-3 Molecular Formula: BH3O3 Molecular Weight (g/mol): 61.83 MDL Number: MFCD00011337 InChI Key: KGBXLFKZBHKPEV-UHFFFAOYSA-N Synonym: orthoboric acid,boracic acid,borofax,boron hydroxide,boron trihydroxide,three elephant,basilit b,trihydroxyborone,h3bo3,flea prufe PubChem CID: 7628 ChEBI: CHEBI:33118 IUPAC Name: boric acid SMILES: OB(O)O
Boric Acid (Crystalline/Electrophoresis), Fisher BioReagents
CAS: 10043-35-3 Molecular Formula: BH3O3 Molecular Weight (g/mol): 61.83 MDL Number: MFCD00011337 InChI Key: KGBXLFKZBHKPEV-UHFFFAOYSA-N Synonym: orthoboric acid,boracic acid,borofax,boron hydroxide,boron trihydroxide,three elephant,basilit b,trihydroxyborone,h3bo3,flea prufe PubChem CID: 7628 ChEBI: CHEBI:33118 IUPAC Name: boric acid SMILES: OB(O)O
Boric acid, 99.99% (metals basis)
CAS: 10043-35-3 Molecular Formula: BH3O3 Molecular Weight (g/mol): 61.83 MDL Number: MFCD00011337 InChI Key: KGBXLFKZBHKPEV-UHFFFAOYSA-N Synonym: orthoboric acid,boracic acid,borofax,boron hydroxide,boron trihydroxide,three elephant,basilit b,trihydroxyborone,h3bo3,flea prufe PubChem CID: 7628 ChEBI: CHEBI:33118 IUPAC Name: boric acid SMILES: OB(O)O
Boric acid, 99.99%, (trace metal basis), extra pure
CAS: 10043-35-3 Molecular Formula: BH3O3 Molecular Weight (g/mol): 61.83 MDL Number: MFCD00011337 InChI Key: KGBXLFKZBHKPEV-UHFFFAOYSA-N Synonym: orthoboric acid,boracic acid,borofax,boron hydroxide,boron trihydroxide,three elephant,basilit b,trihydroxyborone,h3bo3,flea prufe PubChem CID: 7628 ChEBI: CHEBI:33118 IUPAC Name: boric acid SMILES: OB(O)O
Boric Acid, 4% (w/v) with Indicator (Bromocresol Green-Methyl Red), Certified, 4.0% (w/v) ±0.2%, LabChem™
CAS: 10043-35-3 Molecular Formula: BH3O3 Molecular Weight (g/mol): 61.83 MDL Number: MFCD00011337 InChI Key: KGBXLFKZBHKPEV-UHFFFAOYSA-N Synonym: orthoboric acid,boracic acid,borofax,boron hydroxide,boron trihydroxide,three elephant,basilit b,Tris-borate-EDTA,h3bo3,flea prufe PubChem CID: 7628 ChEBI: CHEBI:33118 IUPAC Name: boric acid SMILES: OB(O)O
Boric acid, 99.8%
CAS: 10043-35-3 Molecular Formula: BH3O3 Molecular Weight (g/mol): 61.83 MDL Number: MFCD00011337 InChI Key: KGBXLFKZBHKPEV-UHFFFAOYSA-N Synonym: orthoboric acid,boracic acid,borofax,boron hydroxide,boron trihydroxide,three elephant,basilit b,trihydroxyborone,h3bo3,flea prufe PubChem CID: 7628 ChEBI: CHEBI:33118 IUPAC Name: boric acid SMILES: OB(O)O
Boric acid, ACS, 99.5% min
CAS: 10043-35-3 Molecular Formula: BH3O3 Molecular Weight (g/mol): 61.83 MDL Number: MFCD00011337 InChI Key: KGBXLFKZBHKPEV-UHFFFAOYSA-N Synonym: orthoboric acid,boracic acid,borofax,boron hydroxide,boron trihydroxide,three elephant,basilit b,trihydroxyborone,h3bo3,flea prufe PubChem CID: 7628 ChEBI: CHEBI:33118 IUPAC Name: boric acid SMILES: OB(O)O
Boric acid, 99.5%, for analysis
CAS: 10043-35-3 Molecular Formula: BH3O3 Molecular Weight (g/mol): 61.83 MDL Number: MFCD00011337 InChI Key: KGBXLFKZBHKPEV-UHFFFAOYSA-N Synonym: orthoboric acid,boracic acid,borofax,boron hydroxide,boron trihydroxide,three elephant,basilit b,trihydroxyborone,h3bo3,flea prufe PubChem CID: 7628 ChEBI: CHEBI:33118 IUPAC Name: boric acid SMILES: OB(O)O