accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1004548-96-2

OONH2NOFFFHO

CAS RN 1004548-96-2

IUPAC Name: 6-(2-aminoethyl)-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraen-4-one; trifluoroacetic acid
Synonyms: 6-(2-aminoethyl)-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraen-4-one; trifluoroacetic acid
Molecular Weight (g/mol): 398.38
Molecular Formula: C19H21F3N2O4
InChi Key: XHUSAIWAUAMZRG-UHFFFAOYSA-N
SMILES: OC(=O)C(F)(F)F.NCCC1=CC(=O)OC2=C3CCCN4CCCC(C=C12)=C34

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results