accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOSH

CAS RN 10047-28-6

IUPAC Name: butyl 2-sulfanylacetate
Synonyms: butyl thioglycolate
Molecular Weight (g/mol): 148.22
Molecular Formula: C6H12O2S
InChi Key: SKGVGRLWZVRZDC-UHFFFAOYSA-N
SMILES: CCCCOC(=O)CS
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results