CAS RN 10049-01-1
IUPAC Name:
bismuth(3+) phosphoric acid
Synonyms:
bismuth(3+) phosphoric acid
Molecular Weight (g/mol):
306.97
Molecular Formula:
BiH3O4P
InChi Key:
SFOQXWSZZPWNCL-UHFFFAOYSA-N
SMILES:
[Bi+3].OP(O)(O)=O
1
–
2
of
2
results
1
Bismuth(III) phosphate, 99%
CAS: 10049-01-1 Molecular Formula: BiH3O4P Molecular Weight (g/mol): 306.97 MDL Number: MFCD00049721 InChI Key: SFOQXWSZZPWNCL-UHFFFAOYSA-N Synonym: bismuth phosphate,bismuth iii phosphate,bismugel,phosphoric acid, bismuth 3+ salt 1:1,bismuth phosphate mi,bismuth phosphate who-dd,acmc-20allc,bismuth orthophosphate,bi.po4,bismuth phosphate, PubChem CID: 9839429 SMILES: [Bi+3].OP(O)(O)=O
2
Sigma Aldrich Platinum(0)-1,3-Divinyl-1,1,3,3-Tetramethyldisiloxane Complex Solution
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.