CAS RN 10049-10-2
IUPAC Name:
λ²-chromium(2+) difluoride
Synonyms:
chromium(II) fluoride
Molecular Weight (g/mol):
89.99
Molecular Formula:
CrF2
InChi Key:
RNFYGEKNFJULJY-UHFFFAOYSA-L
SMILES:
[F-].[F-].[Cr++]
1
–
1
of
1
results
1
Chromium(II) fluoride, 95%, Thermo Scientific™
CAS: 10049-10-2 MDL Number: MFCD00016044