accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10049-12-4

IUPAC Name: vanadium(3+) trifluoride
Synonyms: vanadium(III) fluoride
Molecular Weight (g/mol): 107.94
Molecular Formula: F3V
InChi Key: PASVSMBJEYQYSN-UHFFFAOYSA-K
SMILES: [F-].[F-].[F-].[V+3]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results