accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCH3NNH2O

CAS RN 1005-24-9

IUPAC Name: 3-carbamoyl-1-methylpyridin-1-ium chloride
Synonyms: methyl niacinamide chloride
Molecular Weight (g/mol): 172.61
Molecular Formula: C7H9ClN2O
InChi Key: BWVDQVQUNNBTLK-UHFFFAOYSA-N
SMILES: [Cl-].C[N+]1=CC=CC(=C1)C(N)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results