CAS RN 1005-28-3
IUPAC Name:
2-(dimethylamino)-1,3-thiazole-5-carbaldehyde
Synonyms:
2-(dimethylamino)-1,3-thiazole-5-carbaldehyde
Molecular Weight (g/mol):
156.2
Molecular Formula:
C6H8N2OS
InChi Key:
ZOPJEHPVWLTDJW-UHFFFAOYSA-N
SMILES:
CN(C)C1=NC=C(S1)C=O
1
–
1
of
1
results
1
Promotions Available
2-(Dimethylamino)thiazole-5-carboxaldehyde, 97%
2-(Dimethylamino)thiazole-5-carboxaldehyde, 97%
CAS: 1005-28-3 InChI Key: ZOPJEHPVWLTDJW-UHFFFAOYSA-N Synonym: 2-dimethylamino-1,3-thiazole-5-carbaldehyde,2-dimethylamino thiazole-5-carbaldehyde,2-dimethylamino thiazole-5-carboxaldehyde,2-dimethylamino-thiazole-5-carbaldehyde,5-thiazolecarboxaldehyde,2-dimethylamino,5-thiazolecarboxaldehyde, 2-dimethylamino,2-dimethylaminothiazole-5-carbaldehyde PubChem CID: 12846795 IUPAC Name: 2-(dimethylamino)-1,3-thiazole-5-carbaldehyde SMILES: CN(C)C1=NC=C(S1)C=O